Learn More
Selleck Chemical LLC 1, 10-Phenanthroline monohydrate 25mg 5144-89-8 Phenanthroline monohydrate
SDP
Supplier:  Selleck Chemical LLC S554325MG
Encompass_Preferred
1,10-Phenanthroline is a classic chelating bidentate ligand for transition metal ions that has played an important role in the development of coordination chemistry. It is an inhibitor of metallopeptidases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8270