Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,10-Phenanthroline monohydrate ACS reagent, puriss. p.a., >=99.5% (calc. to the dried substance), for redox titration | 5144-89-8 | MFCD00149973 | 5G
Catalog No. 501766953
Encompass
1,10-Phenanthroline monohydrate ACS reagent, puriss. p.a., >=99.5% (calc. to the dried substance), for redox titration | Purity: >=99.5% (calc. to the dried substance); 8.5-9.5% water basis | Mol Wt: 198.22 | 5144-89-8 | MFCD00149973 | 5G
Catalog No. 50-176-6953 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 335105G
1,10-Phenanthroline monohydrate ACS reagent, puriss. p.a., >=99.5% (calc. to the dried substance), for redox titration | Purity: >=99.5% (calc. to the dried substance); 8.5-9.5% water basis | Mol Wt: 198.22 | 5144-89-8 | MFCD00149973 | 5G