Learn More
Medchemexpress LLC 1,10-phenanthroline monohydrate | 5144-89-8 | MFCD00149973 | 1mL
SDP
Catalog No. 5000266340
Encompass_Preferred
o-Phenanthroline (1 10-Phenanthroline) monohydrate a metal chelator prevents the induction of chromosomal aberrations in streptozotocin-treated cells o-Phenanthroline monohydrate forms a red chelate with Fe2 that absorbs maximally at 510 nm o-Phenanthroline (1 10-Phenanthroline) monohydrate is also a MMP inhibitor[1][2]
Catalog No. 50-002-66340 Supplier Medchemexpress LLC Supplier No. HYY184110MM/1ML
Add to Cart
Add to Cart
o-Phenanthroline (1 10-Phenanthroline) monohydrate a metal chelator prevents the induction of chromosomal aberrations in streptozotocin-treated cells o-Phenanthroline monohydrate forms a red chelate with Fe2 that absorbs maximally at 510 nm o-Phenanthroline (1 10-Phenanthroline) monohydrate is also a MMP inhibitor[1][2]