Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,10-Phenanthroline monohydrochloride monohydrate

Catalog No. P1300210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1300210G 10 g
P1300225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1300210G Supplier Sigma Aldrich Supplier No. P1300210G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,10-Phenanthroline monohydrochloride monohydrate

Specifications

CAS 18851-33-7
Melting Point 224°C to 225°C (lit.)
Molecular Formula C12H8N2 -+ HCl -+ H2O
Linear Formula C12H8N2 -+ HCl -+ H2O
MDL Number MFCD00150061
Quantity 10 g
Synonym 1,10-Phenanthroline hydrochloride monohydrate; o-Phenanthroline hydrochloride monohydrate
Molecular Weight (g/mol) 234.68
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.