Learn More
Cayman Chemical 1,10-Phenanthroline (hydrate), Purity: ≥98%, CAS: 5144-89-8, 100 g
SDP
Catalog No. 502940949
Encompass_Preferred
A metal chelator and inhibitor of metalloproteases; inhibits zinc-dependent hydrolysis of Fa-Gly-Leu-NH2 by B. subtilis neutral protease and B. thermoproteolyticus thermolysin; fungistatic against P. verrucosa (MIC = 0.8 μg/ml); inhibits zinc-induced P. verrucosa metallo-type peptidase activity; induces uncoupling of and inhibits egg production by adult S. mansoni worm pairs from 0.5-150 μM; reduces worm burden in a mouse model of S. mansoni infection at 20 mg/kg per day
Catalog No. 50-294-0949 Supplier Cayman Chemical Supplier No. 28951100 G
A metal chelator and inhibitor of metalloproteases; inhibits zinc-dependent hydrolysis of Fa-Gly-Leu-NH2 by B. subtilis neutral protease and B. thermoproteolyticus thermolysin; fungistatic against P. verrucosa (MIC = 0.8 μg/ml); inhibits zinc-induced P. verrucosa metallo-type peptidase activity; induces uncoupling of and inhibits egg production by adult S. mansoni worm pairs from 0.5-150 μM; reduces worm burden in a mouse model of S. mansoni infection at 20 mg/kg per day