Learn More
USA Scientific Inc 100 ul round gel filter tip, racks, sterile
Catalog No. NC9849187
Encompass
100 ul Gel-Saver round gel loading filter tip, 0.6 mm OD orifice, sterilized, 5 racks of 204 (1020 tips)
Catalog No. NC9849187 Supplier USA Scientific Inc Supplier No. 10220810
100 ul Gel-Saver round gel loading filter tip, 0.6 mm OD orifice, sterilized, 5 racks of 204 (1020 tips)