Learn More
Thermco Products Inc Accu-Safe Non-Mercury Laboratory Thermometer
Catalog No. NC1810154
Encompass_Preferred
Accu-Safe Non-Mercury Laboratory Thermometer | -1 to 101°C | 0.2°C Div | 450mm Length
Catalog No. NC1810154 Supplier Thermco Products Inc Supplier No. ACC10053BLS
Accu-Safe Non-Mercury Laboratory Thermometer | -1 to 101°C | 0.2°C Div | 450mm Length