Learn More
Medchemexpress LLC 1,11-bis-maleimidotetraethyleneglycol | 86099-06-1 | 98.0% | 25 MG
SDP
Catalog No. 5000341431
Encompass_Preferred
BM-PEG3 is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) comprise two distinct ligands joined by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These chimeras leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Features two distinct ligands connected by a linker
  • Ligands target an E3 ubiquitin ligase and a target protein
  • Employs the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-41431 Supplier Medchemexpress LLC Supplier No. 25MGHY130898
BM-PEG3 is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) comprise two distinct ligands joined by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These chimeras leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is intended for research use only.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Features two distinct ligands connected by a linker
  • Ligands target an E3 ubiquitin ligase and a target protein
  • Employs the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins