Learn More
Medchemexpress LLC 1,11-Bis-maleimidotetraethyleneglycol | 86099-06-1 | 98.0% | 250 MG
SDP
Catalog No. 5000341463
Encompass_Preferred
BM-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands
  • One ligand for E3 ubiquitin ligase
  • Other ligand for target protein
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41463 Supplier Medchemexpress LLC Supplier No. 250MGHY130898
BM-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands
  • One ligand for E3 ubiquitin ligase
  • Other ligand for target protein
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins