Learn More
Medchemexpress LLC 1,11-Bis-maleimidotetraethyleneglycol | 86099-06-1 | 98.0% | 50 MG
SDP
Catalog No. 5000338478
Encompass_Preferred
BM-PEG3 is a polyethylene glycol (PEG)-based linker designed for use in the synthesis of PROTACs (proteolysis-targeting chimeras). PROTACs are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound has a molecular weight of 352.34 and a chemical formula of C16H20N2O7, appearing as a white to light yellow solid. It is soluble in DMSO, with recommendations for proper storage and handling for both powder and solvent forms.
  • It is a polyethylene glycol-based linker.
  • It is used in the synthesis of proteolysis-targeting chimeras.
  • It aids in the degradation of target proteins.
  • It has a high purity of 98.0%.
  • It is available as a white to light yellow solid.
  • It can be dissolved in DMSO for in vitro experiments.
  • It can be formulated for in vivo studies using specific co-solvents like DMSO and corn oil.
Catalog No. 50-003-38478 Supplier Medchemexpress LLC Supplier No. HY13089850MG
BM-PEG3 is a polyethylene glycol (PEG)-based linker designed for use in the synthesis of PROTACs (proteolysis-targeting chimeras). PROTACs are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound has a molecular weight of 352.34 and a chemical formula of C16H20N2O7, appearing as a white to light yellow solid. It is soluble in DMSO, with recommendations for proper storage and handling for both powder and solvent forms.
  • It is a polyethylene glycol-based linker.
  • It is used in the synthesis of proteolysis-targeting chimeras.
  • It aids in the degradation of target proteins.
  • It has a high purity of 98.0%.
  • It is available as a white to light yellow solid.
  • It can be dissolved in DMSO for in vitro experiments.
  • It can be formulated for in vivo studies using specific co-solvents like DMSO and corn oil.