Learn More
Medchemexpress LLC 1,11-Bis(methanesulfonyloxy)-3,6,9-trioxandecane | 55400-73-2 | 95.0% | 350.41 | 1 G
SDP
Catalog No. 5000403577
Encompass_Preferred
Ms-PEG4-MS is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-03577 Supplier Medchemexpress LLC Supplier No. HY427741G
Ms-PEG4-MS is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins