Learn More
Medchemexpress LLC 1,11-Bis(methanesulfonyloxy)-3,6,9-trioxandecane | 55400-73-2 | 95.0% | 350.41 | 5 G
SDP
Catalog No. 5000404659
Encompass_Preferred
Ms-PEG4-MS is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for a target protein.
  • PEG-based linker for PROTAC synthesis
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
  • Available in various quantities for research needs
Catalog No. 50-004-04659 Supplier Medchemexpress LLC Supplier No. HY427745G
Ms-PEG4-MS is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for a target protein.
  • PEG-based linker for PROTAC synthesis
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
  • Available in various quantities for research needs