Learn More
Labrepco Inc (1) 11 CANISTER FOR CXR100 SH
SDP
Catalog No. 509020754
Encompass_Preferred
Canister; Labrepco; For CXR100 Cryo shipper; Prevents liquid spill; New rugged construction; Complies with IATA regulation; 11in.
Catalog No. 50-902-0754 Supplier Labrepco Inc Supplier No. CXR1009C24
Canister; Labrepco; For CXR100 Cryo shipper; Prevents liquid spill; New rugged construction; Complies with IATA regulation; 11in.