Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,11-Diamino-3,6,9-Trioxaundecane

Catalog No. 40421100MGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-421-100MGF 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 40-421-100MGF Supplier Sigma Aldrich Supplier No. 40421100MGF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydrophilic, homo-bifunctional linker, i.e. for the prep. of biotinylation reagents, for the prep. of dendrimers and for the prep. of macrocycles.

Specifications

CAS 162578-86-1
Density 0.86 g/mL at 25°C
Boiling Point 140°C to 170°C (0.05 mmHg, lit.)
Molecular Formula C26H55ClSi
Linear Formula CH3(CH2)17Si[CH2CH(CH3)2]2Cl
MDL Number MFCD00191911
Quantity 5 mL
Synonym 2,2'-[Oxybis(ethylenoxy)]diethylamine
Molecular Weight (g/mol) 431.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.