Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,11-Diazido-3,6,9-Trioxaundecane

Catalog No. 7659531G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-595-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 76-595-31G Supplier Sigma Aldrich Supplier No. 7659531G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,11-Diazido-3,6,9-trioxaundecane may be used to synthesize: 1,11-bis[4-(pyren-1-ylmethoxymethyl)-1H-1,2,3-triazole-1-yl]-3,6,9-trioxaundecane, a fluorogenic chemosensor that can selective detect Hg2+ and Ag+ ions in aqueous methanol solution1-amino-11-azido-3,6,9-trioxaundecane, an intermediate to synthesize tetraethylene glycol based bidentate ligand functionalized with dihydrolipoic acid and biotin (DHLA-TEG-biotin)

Specifications

CAS 762245-84-1
Melting Point 216°C to 221°C
Molecular Formula C21H43BrN3O2
Refractive Index n20/D 1.470
Linear Formula C21H43BrN3O2
Quantity 1 g
Synonym WS-TEMPO; water soluble N-oxyl compound; water soluble-TEMPO
Molecular Weight (g/mol) 449.49 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.