Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,11-Dodecadiene 99.0+%, TCI America™
SDP

Catalog No. D02835ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02835ML 5 mL
D028325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D02835ML Supplier TCI America Supplier No. D02835ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5876-87-9
Molecular Formula C12H22
Molecular Weight (g/mol) 166.308
MDL Number MFCD00048156
InChI Key IYPLTVKTLDQUGG-UHFFFAOYSA-N
PubChem CID 22170
IUPAC Name dodeca-1,11-diene
SMILES C=CCCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 207°C
Quantity 5 mL
Formula Weight 166.31
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,11-Dodecadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.