Learn More
Selleck Chemical LLC 1 11-Undecanedicarboxylic acid
SDP
Catalog No. 5000003768
Encompass_Preferred
1 11-Undecanedicarboxylic acid (Tridecanedioic acid Brassylic acid Brassilic acid) is an unusual odd-numbered dicarboxylic acid that appears in the urines of children with neonatal adrenoleukodystrophy and Zellweger syndrome as an additional marker of these peroxisomal disorders
Catalog No. 50-000-03768 Supplier Selleck Chemical LLC Supplier No. S606325MG
1 11-Undecanedicarboxylic acid (Tridecanedioic acid Brassylic acid Brassilic acid) is an unusual odd-numbered dicarboxylic acid that appears in the urines of children with neonatal adrenoleukodystrophy and Zellweger syndrome as an additional marker of these peroxisomal disorders