Learn More
Medchemexpress LLC 1,11b-Dihydro-11b-hydroxymaackiain | 210537-05-6 | 302.28 | 1 MG
SDP
Catalog No. 5000417069
Encompass_Preferred
1,11b-Dihydro-11b-hydroxymaackiain is a pterocarpane extract from the stems of *Erycibe expansa*, which shows a hepatoprotective effect. It is for research use only, and not sold to patients.
  • Hepatoprotective effect
  • Purity: 97%
Catalog No. 50-004-17069 Supplier Medchemexpress LLC Supplier No. HYN09751MG
1,11b-Dihydro-11b-hydroxymaackiain is a pterocarpane extract from the stems of *Erycibe expansa*, which shows a hepatoprotective effect. It is for research use only, and not sold to patients.
  • Hepatoprotective effect
  • Purity: 97%