Learn More
Medchemexpress LLC 1,12-dodecanediol | 5675-51-4 | MFCD00004755 | 99.2% | 202.34 g/mol | C12H26O2 | 100 G
SDP
Catalog No. 5000303788
Encompass_Preferred
Dodecane-1,12-diol (1,12-dodecanediol) is a linear aliphatic diol used as a biochemical reagent and intermediate in organic and polymer synthesis. It serves as a building block in enzymatic synthesis of bio-based polyesters and is used in formulations and research where a long-chain diol is required. The compound is a solid with formula C12H26O2 and a molecular weight of about 202.34 g/mol.
  • Used in enzymatic synthesis of bio-based polyesters.
  • Serves as a building block for surfactants, lubricants, and plasticizers.
  • High purity suitable for research and synthetic applications.
  • Available in multiple package sizes including 100 g for lab use.
  • Solid at room temperature; follow SDS for safe handling and storage.
Catalog No. 50-003-03788 Supplier Medchemexpress LLC Supplier No. HYW014779100G
Dodecane-1,12-diol (1,12-dodecanediol) is a linear aliphatic diol used as a biochemical reagent and intermediate in organic and polymer synthesis. It serves as a building block in enzymatic synthesis of bio-based polyesters and is used in formulations and research where a long-chain diol is required. The compound is a solid with formula C12H26O2 and a molecular weight of about 202.34 g/mol.
  • Used in enzymatic synthesis of bio-based polyesters.
  • Serves as a building block for surfactants, lubricants, and plasticizers.
  • High purity suitable for research and synthetic applications.
  • Available in multiple package sizes including 100 g for lab use.
  • Solid at room temperature; follow SDS for safe handling and storage.