Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,13-Tetradecadiene, 90+%

Catalog No. AAH5347906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5347906 5 g
AAH5347903 1 g
AAH5347914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5347906 Supplier Thermo Scientific Chemicals Supplier No. H5347906
Only null left
Add to Cart

CAS: 21964-49-8 | C14H26 | 194.362 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21964-49-8
Molecular Formula C14H26
Molecular Weight (g/mol) 194.362
MDL Number MFCD00014941
InChI Key XMRSTLBCBDIKFI-UHFFFAOYSA-N
Synonym 1,13-tetradecadiene, tetradecadiene-1,13, acmc-209fpz
PubChem CID 30875
IUPAC Name tetradeca-1,13-diene
SMILES C=CCCCCCCCCCCC=C

Specifications

Density 0.849
Boiling Point 131°C (17 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.443
Linear Formula CH2=CH(CH2)10CH=CH2
Quantity 5 g
Beilstein 1741459
Formula Weight 194.36
Percent Purity ≥90%
Chemical Name or Material 1,13-Tetradecadiene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-687-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.