Learn More
Waters Corp Ferrule, 1/16 in., VG2 Vespel-Graphite
Catalog No. 502392548 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a 1/16 in. VG2 Vespel-Graphite ferrule, designed for use with Xevo TQ-GC Mass Spectrometers.
  • Ferrule for precise connections
  • Made from VG2 Vespel-Graphite material
  • 1/16 in. size for specific applications
  • Compatible with Xevo TQ-GC Mass Spectrometers
Catalog No. 50-239-2548 Supplier Waters Corp Supplier No. 700011902
This product is a 1/16 in. VG2 Vespel-Graphite ferrule, designed for use with Xevo TQ-GC Mass Spectrometers.
  • Ferrule for precise connections
  • Made from VG2 Vespel-Graphite material
  • 1/16 in. size for specific applications
  • Compatible with Xevo TQ-GC Mass Spectrometers