Learn More
Waters Corp Waters Tubing, PEEK, 0.004 in. ID, 60 in. Length
Catalog No. NC9866504 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PEEK tubing is designed for UPLC systems, offering precise fluid transfer in analytical applications. The black color of the tubing may also provide light-blocking properties for sensitive compounds.
  • Designed for UPLC systems.
  • Features a 0.004 in. (0.1016 mm) internal diameter and 0.0625 in. (1.5875 mm) external diameter for precise fluid transfer.
  • Black color may provide light-blocking properties for sensitive compounds.
  • Ensures continued operation and maintenance of UPLC systems to minimize downtime.
Catalog No. NC9866504 Supplier Waters Corp Supplier No. 700003799
This PEEK tubing is designed for UPLC systems, offering precise fluid transfer in analytical applications. The black color of the tubing may also provide light-blocking properties for sensitive compounds.
  • Designed for UPLC systems.
  • Features a 0.004 in. (0.1016 mm) internal diameter and 0.0625 in. (1.5875 mm) external diameter for precise fluid transfer.
  • Black color may provide light-blocking properties for sensitive compounds.
  • Ensures continued operation and maintenance of UPLC systems to minimize downtime.