Learn More
Optimize Technologies 1/16" OD x .010" ID x 50ft ETFE Tubing
SDP
Catalog No. 502006193
Encompass_Preferred
Optimize's ETFE tubing maintains excellent chemical resistance, withstands aqueous pressures up to 4,000psi and is available in both 1/8" and 1/16" Odds. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.
Catalog No. 50-200-6193 Supplier Optimize Technologies Supplier No. 101400194
Optimize's ETFE tubing maintains excellent chemical resistance, withstands aqueous pressures up to 4,000psi and is available in both 1/8" and 1/16" Odds. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.