Learn More
Optimize Technologies 1/16" OD x .020" ID x 10ft ETFE Tubing
SDP
Catalog No. 502006194
Encompass_Preferred
Optimize's ETFE tubing maintains excellent chemical resistance, withstands aqueous pressures up to 4,000psi and is available in both 1/8" and 1/16" Ods. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.
Catalog No. 50-200-6194 Supplier Optimize Technologies Supplier No. 101400196
Optimize's ETFE tubing maintains excellent chemical resistance, withstands aqueous pressures up to 4,000psi and is available in both 1/8" and 1/16" Ods. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.