Learn More
Optimize Technologies 1/16" OD x .030" ID x 10ft PEEK Tubing
SDP
Catalog No. 50113212
Encompass_Preferred
Optimize's PEEK tubing is biocompatible and chemically inert to most commonly-used solvents and can withstand pressures up to 6,000 psi. It remains a great alternative to stainless steel tubing for high pressure applications, withstands continuous use at HPLC pressure without swelling or bursting, its not affected by aggressive mobile phases and will not corrode. The tubing is available in many different lengths and OD/ID sizes. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.
Catalog No. 50-113-212 Supplier Optimize Technologies Supplier No. 101300187
Optimize's PEEK tubing is biocompatible and chemically inert to most commonly-used solvents and can withstand pressures up to 6,000 psi. It remains a great alternative to stainless steel tubing for high pressure applications, withstands continuous use at HPLC pressure without swelling or bursting, its not affected by aggressive mobile phases and will not corrode. The tubing is available in many different lengths and OD/ID sizes. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.