Learn More
Optimize Technologies 1/16" OD x .030" ID x 50ft ETFE Tubing
SDP
Catalog No. 50113219
Encompass_Preferred
Optimize's ETFE tubing maintains excellent chemical resistance, withstands aqueous pressures up to 4,000psi and is available in both 1/8" and 1/16" Ods. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.
Catalog No. 50-113-219 Supplier Optimize Technologies Supplier No. 101400200
Optimize's ETFE tubing maintains excellent chemical resistance, withstands aqueous pressures up to 4,000psi and is available in both 1/8" and 1/16" Ods. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.