Learn More
Optimize Technologies 1/16" OD x .040" ID x 10ft PTFE Tubing, Natural
SDP
Catalog No. 50113313
Encompass_Preferred
Optimize's PTFE tubing is chemically inert to most solvents, easy to cut, translucent for easy monitoring and recommended for general low pressure applications. The tubing is also available in various OD and ID sizes. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.
Catalog No. 50-113-313 Supplier Optimize Technologies Supplier No. 101500232
Optimize's PTFE tubing is chemically inert to most solvents, easy to cut, translucent for easy monitoring and recommended for general low pressure applications. The tubing is also available in various OD and ID sizes. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.