Learn More
SWAGELOK TEXAS MID COAST 1/16 T. X 1/8 TX REDUCER
Catalog No. NC2306767
Encompass
Stainless Steel Swagelok Tube Fitting, Reducer, 1/16 in. x 1/8 in. Tube OD
Catalog No. NC2306767 Supplier SWAGELOK TEXAS MID COAST Supplier No. SS100R2
Stainless Steel Swagelok Tube Fitting, Reducer, 1/16 in. x 1/8 in. Tube OD