Learn More
Glass Expansion 1/16" Tefzel Ferrules (PKT 10)
SDP
Catalog No. 502703063
Encompass_Preferred
1.6mm Tefzel Ferrules (PKT 10). Package of ten replacement flangeless Tefzel ferrules 1.6mm (1/16”). Compatible with Glass Expansion's High Pressure Nebulizer Interface Kits (FT-16-8-X and FT-16-8). For 1/8” Tefzel Ferrules see 70-803-1513. Note: these 1/8” ferrules are only for use with FT-16-8-X. For FT-16-8 use 1/8” Tefzel Flangeless Ferrule 70-803-1316.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3063 Supplier Glass Expansion Supplier No. 708031315
1.6mm Tefzel Ferrules (PKT 10). Package of ten replacement flangeless Tefzel ferrules 1.6mm (1/16”). Compatible with Glass Expansion's High Pressure Nebulizer Interface Kits (FT-16-8-X and FT-16-8). For 1/8” Tefzel Ferrules see 70-803-1513. Note: these 1/8” ferrules are only for use with FT-16-8-X. For FT-16-8 use 1/8” Tefzel Flangeless Ferrule 70-803-1316.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001