Learn More
Waters Corp Tubing, PEEK, 1/16 in. x 0.007 in., 1 m
Catalog No. 50792290 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PEEK tubing is designed for various laboratory applications. It features precise dimensions of 1/16 in. outer diameter and 0.007 in. inner diameter, with a length of 1 m. This tubing is suitable for use in HPLC and MS systems.
  • Constructed from PEEK material
  • Outer diameter of 1/16 in.
  • Inner diameter of 0.007 in.
  • Length of 1 m
  • Yellow in color
  • Sold in 1-pack units
  • Compatible with HPLC and MS system types
  • Suitable for ACQUITY TQ Detector and SQ Detector instruments
  • Replacement is recommended as required
Catalog No. 50-792-290 Supplier Waters Corp Supplier No. 6436004S1
This PEEK tubing is designed for various laboratory applications. It features precise dimensions of 1/16 in. outer diameter and 0.007 in. inner diameter, with a length of 1 m. This tubing is suitable for use in HPLC and MS systems.
  • Constructed from PEEK material
  • Outer diameter of 1/16 in.
  • Inner diameter of 0.007 in.
  • Length of 1 m
  • Yellow in color
  • Sold in 1-pack units
  • Compatible with HPLC and MS system types
  • Suitable for ACQUITY TQ Detector and SQ Detector instruments
  • Replacement is recommended as required