Learn More
Optimize Technologies 1/16" OD x .030" ID x 10cm Stainless Steel Tubing
SDP
Catalog No. 50113303
Encompass_Preferred
Optimize's stainless steel tubing is pre-cut, micro polished, seamless, guarantees square/burr-free ends, and is passivated prior to shipment. The tubing is available in many different lengths and OD/ID sizes. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.
Catalog No. 50-113-303 Supplier Optimize Technologies Supplier No. 101200160
Optimize's stainless steel tubing is pre-cut, micro polished, seamless, guarantees square/burr-free ends, and is passivated prior to shipment. The tubing is available in many different lengths and OD/ID sizes. Please contact your Fisher representative or Optimize directly to view a full list of tubing options.