Learn More
Ace Glass, Inc. PTFE 1/16in tube compression fitting to 1/8in male NPT
SDP
Catalog No. 50977668
Encompass
1/16 PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-668 Supplier Ace Glass, Inc. Supplier No. 1270902
1/16 PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.