Learn More
CELLTREAT Scientific Products 1.1mL Bacteriological/Milk Pipet, Bulk Packed in Bags, Sterile
SDP
Catalog No. NC3433451
Encompass_Preferred
"Bacteriological/Milk pipets are manufactured to meet APHA requirements shown in Standard Methods for Examination of Dairy Products and ASTM E1043-00(2010) Standard Specification for Pipets, Milk and Cream ExaminationSpecialized graduations printed in bold easy-to-ready black ink allow quick volume identificationPremium polymer fiber barrier plug outperforms cotton plugs"
Catalog No. NC3433451 Supplier CELLTREAT Scientific Products Supplier No. 229257
"Bacteriological/Milk pipets are manufactured to meet APHA requirements shown in Standard Methods for Examination of Dairy Products and ASTM E1043-00(2010) Standard Specification for Pipets, Milk and Cream ExaminationSpecialized graduations printed in bold easy-to-ready black ink allow quick volume identificationPremium polymer fiber barrier plug outperforms cotton plugs"