Learn More
SAI Infusion Technologies 1.1ml Serum Gel Blood Tubes with Flip Cap
SDP
Catalog No. NC1029089
Encompass
1.1ml Serum Gel Blood Tube with Clotting Activator, Serum Separator Gel and attached flip cap. Ideal for collection of up to 1.1ml of blood for serum. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only.
Catalog No. NC1029089 Supplier SAI Infusion Technologies Supplier No. PMTPSG1.1
1.1ml Serum Gel Blood Tube with Clotting Activator, Serum Separator Gel and attached flip cap. Ideal for collection of up to 1.1ml of blood for serum. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only.