Learn More
MSE Supplies LLC 1.1MM THICK UNCOATED 100X100MM
SDP
Catalog No. NC1899912
Encompass
1.1 mm Thick Uncoated Soda Lime Glass Substrates, >90% Transmittance.100 mm x 100 mm x 1.1mm
Catalog No. NC1899912 Supplier MSE Supplies LLC Supplier No. GL0607
1.1 mm Thick Uncoated Soda Lime Glass Substrates, >90% Transmittance.100 mm x 100 mm x 1.1mm