Learn More
MSC Gibraltar P14641GB 2,400 Lb Capacity, Steel, 1/2-13 Thread, Fixed Lifting Eye Bolt Fully Threaded, 1-1/2" Shank, 1-1/2" Thread Length, Shoulder
Catalog No. 501110520
Encompass
Gibraltar P14641GB 2,400 Lb Capacity, Steel, 1/2-13 Thread, Fixed Lifting Eye Bolt Fully Threaded, 1-1/2" Shank, 1-1/2" Thread Length, Shoulder
Catalog No. 50-111-0520 Supplier MSC Supplier No. 73155038
Gibraltar P14641GB 2,400 Lb Capacity, Steel, 1/2-13 Thread, Fixed Lifting Eye Bolt Fully Threaded, 1-1/2" Shank, 1-1/2" Thread Length, Shoulder