Learn More
Grainger Chicago-Latrobe Hand Tap Set, 1/2in.-20 UNF, H3, 3-Piece Set
Catalog No. 502753865
Encompass_Preferred
This three-piece hand tap set is designed for manual threading operations, offering precise control at lower speeds. Constructed from high-speed steel, the taps provide excellent wear resistance and flexibility for reliable performance in soft materials, including aluminum and low-carbon steel, as well as hardened steels. The straight-flute design delivers free-cutting action and minimal heat generation, ensuring a smooth finish on the hole.
  • Includes one each taper, plug, and bottoming chamfer
  • High-speed steel construction for durability and shock absorption
  • Bright uncoated finish suitable for resharpening
  • Straight-flute design for efficient chip management in through holes
  • Right-hand thread direction
  • Thread size: 1/2"-20 UNF
  • Class of fit: 3B
  • Thread limit: H3
  • Number of flutes: 4
  • Overall length: 3-3/8 in.
Catalog No. 50-275-3865 Supplier Grainger Supplier No. 15J780
This three-piece hand tap set is designed for manual threading operations, offering precise control at lower speeds. Constructed from high-speed steel, the taps provide excellent wear resistance and flexibility for reliable performance in soft materials, including aluminum and low-carbon steel, as well as hardened steels. The straight-flute design delivers free-cutting action and minimal heat generation, ensuring a smooth finish on the hole.
  • Includes one each taper, plug, and bottoming chamfer
  • High-speed steel construction for durability and shock absorption
  • Bright uncoated finish suitable for resharpening
  • Straight-flute design for efficient chip management in through holes
  • Right-hand thread direction
  • Thread size: 1/2"-20 UNF
  • Class of fit: 3B
  • Thread limit: H3
  • Number of flutes: 4
  • Overall length: 3-3/8 in.