Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,10b-Tetrahydrofluoranthene 98.0+%, TCI America™
SDP

Catalog No. T07715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07715G 5 g
T07711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07715G Supplier TCI America Supplier No. T07715G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20279-21-4
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00045348
InChI Key VMBZUGDYWLFLEU-UHFFFAOYNA-N
PubChem CID 89305
IUPAC Name 1,2,3,10b-tetrahydrofluoranthene
SMILES C1CC2C3=CC=CC=C3C3=CC=CC(C1)=C23

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 205°C
Quantity 5 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,10b-Tetrahydrofluoranthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.