Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,3,4,5,6-hexabromocyclohexane | 1837-91-8 | MFCD00059127 | 98.0% | 557.54 | C6H6Br6 | 50 MG
SDP

Catalog No. 5000240047
Encompass_Preferred

Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane) is a brominated cyclic hydrocarbon supplied as a solid research reagent. It is reported to potently inhibit JAK2 tyrosine kinase autophosphorylation and is provided with supporting documentation for laboratory use.

  • High purity (98.0%).
  • Chemical formula C6H6Br6 and molecular weight 557.54 g/mol.
  • Solid physical form suitable for storage and handling.
  • CAS registry number 1837-91-8 for unequivocal identification.
  • Documentation available: COA, SDS, and data sheet.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-40047 Supplier Medchemexpress LLC Supplier No. HY10759450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane) is a brominated cyclic hydrocarbon supplied as a solid research reagent. It is reported to potently inhibit JAK2 tyrosine kinase autophosphorylation and is provided with supporting documentation for laboratory use.

  • High purity (98.0%).
  • Chemical formula C6H6Br6 and molecular weight 557.54 g/mol.
  • Solid physical form suitable for storage and handling.
  • CAS registry number 1837-91-8 for unequivocal identification.
  • Documentation available: COA, SDS, and data sheet.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.