Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,3,4,5,6-hexabromocyclohexane | 1837-91-8 | MFCD00059127 | 98.0% | 557.54 g/mol | C6H6Br6 | 1 ML
SDP

Catalog No. 5000240689
Encompass_Preferred

Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane; C6H6Br6) is a bromohydrocarbon that inhibits JAK2 tyrosine kinase autophosphorylation. It is supplied for laboratory research in solution and solid formats, commonly as a 10 mM solution in DMSO (1 mL) or as mg-scale solids. Intended for research use only.

  • Potent inhibitor of JAK2 autophosphorylation.
  • Molecular weight 557.54 g/mol.
  • Purity 98.0% by HPLC.
  • Available as 10 mM solution in DMSO (1 mL) and as solid in mg quantities.
  • Suitable for biochemical and cell-based assay research.

Catalog No. 50-002-40689 Supplier Medchemexpress LLC Supplier No. HY10759410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane; C6H6Br6) is a bromohydrocarbon that inhibits JAK2 tyrosine kinase autophosphorylation. It is supplied for laboratory research in solution and solid formats, commonly as a 10 mM solution in DMSO (1 mL) or as mg-scale solids. Intended for research use only.

  • Potent inhibitor of JAK2 autophosphorylation.
  • Molecular weight 557.54 g/mol.
  • Purity 98.0% by HPLC.
  • Available as 10 mM solution in DMSO (1 mL) and as solid in mg quantities.
  • Suitable for biochemical and cell-based assay research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.