Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,3,4,5,6-hexabromocyclohexane | 1837-91-8 | MFCD00059127 | 98.0% | 557.54 g/mol | C6H6Br6 | 10 MG
SDP

Catalog No. 5000240028
Encompass_Preferred

Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane) is a hexabrominated cyclohexane research reagent used in biochemical studies. It is described as a potent inhibitor of JAK2 tyrosine kinase autophosphorylation and is supplied as a solid with high purity for in vitro assays and synthetic applications.

  • High purity 98.0%.
  • Molecular weight 557.54 g/mol.
  • Formula C6H6Br6.
  • Physical state solid, white to off-white.
  • Available in small milligram pack sizes and as 10 mM solutions in DMSO.
  • Reported potent inhibitor of JAK2 tyrosine kinase autophosphorylation.
  • Suitable for in vitro biochemical assays and as a synthetic intermediate.

Catalog No. 50-002-40028 Supplier Medchemexpress LLC Supplier No. HY10759410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane) is a hexabrominated cyclohexane research reagent used in biochemical studies. It is described as a potent inhibitor of JAK2 tyrosine kinase autophosphorylation and is supplied as a solid with high purity for in vitro assays and synthetic applications.

  • High purity 98.0%.
  • Molecular weight 557.54 g/mol.
  • Formula C6H6Br6.
  • Physical state solid, white to off-white.
  • Available in small milligram pack sizes and as 10 mM solutions in DMSO.
  • Reported potent inhibitor of JAK2 tyrosine kinase autophosphorylation.
  • Suitable for in vitro biochemical assays and as a synthetic intermediate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.