Learn More
Medchemexpress LLC 1,2,3,4,5,6-hexabromocyclohexane | 1837-91-8 | MFCD00059127 | 98.0% | 557.54 g·mol⁻¹ | C6H6Br6 | 25 MG
SDP
Catalog No. 5000241458
Encompass_Preferred
Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane) is a brominated small molecule reported to inhibit JAK2 tyrosine kinase autophosphorylation. Supplied as a solid research reagent, it is intended for biochemical and cell-based studies investigating JAK2 signaling and related pathways.
  • Potent inhibitor of JAK2 tyrosine kinase autophosphorylation.
  • Chemical formula C6H6Br6 and molecular weight 557.54 g·mol⁻¹.
  • High reported purity (~98%) suitable for research use.
  • Available as a solid in small research-scale quantities (e.g., 25 mg).
  • Relevant for biochemical and cell-based assay applications.
Catalog No. 50-002-41458 Supplier Medchemexpress LLC Supplier No. HY10759425MG
Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane) is a brominated small molecule reported to inhibit JAK2 tyrosine kinase autophosphorylation. Supplied as a solid research reagent, it is intended for biochemical and cell-based studies investigating JAK2 signaling and related pathways.
  • Potent inhibitor of JAK2 tyrosine kinase autophosphorylation.
  • Chemical formula C6H6Br6 and molecular weight 557.54 g·mol⁻¹.
  • High reported purity (~98%) suitable for research use.
  • Available as a solid in small research-scale quantities (e.g., 25 mg).
  • Relevant for biochemical and cell-based assay applications.