Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,3,4,5,6-hexabromocyclohexane | 1837-91-8 | MFCD00059127 | 98.0% | 557.54 g/mol | C6H6Br6 | 5 MG
SDP

Catalog No. 5000240109
Encompass_Preferred

Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane) is a hexabrominated bromohydrocarbon used as a research reagent. It is reported to inhibit JAK2 tyrosine kinase autophosphorylation and is provided with high purity for biochemical and cell signaling studies.

  • High purity (98.0%) for reliable experimental results.
  • Molecular weight 557.54 g/mol and formula C6H6Br6 for formulation and calculation.
  • Available in small milligram quantities suitable for screening and assay development.
  • Soluble in DMSO, facilitating preparation of stock solutions.
  • Useful as a JAK2 autophosphorylation inhibitor in signaling and kinase assays.

Catalog No. 50-002-40109 Supplier Medchemexpress LLC Supplier No. HY1075945MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzene hexabromide (1,2,3,4,5,6-hexabromocyclohexane) is a hexabrominated bromohydrocarbon used as a research reagent. It is reported to inhibit JAK2 tyrosine kinase autophosphorylation and is provided with high purity for biochemical and cell signaling studies.

  • High purity (98.0%) for reliable experimental results.
  • Molecular weight 557.54 g/mol and formula C6H6Br6 for formulation and calculation.
  • Available in small milligram quantities suitable for screening and assay development.
  • Soluble in DMSO, facilitating preparation of stock solutions.
  • Useful as a JAK2 autophosphorylation inhibitor in signaling and kinase assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.