Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2,3,4,5-Pentamethylcyclopentadiene

Catalog No. 2140275G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-402-75G 5 g
21-402-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-75G Supplier Sigma Aldrich Supplier No. 2140275G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Mechanism of the diels-alder reaction of paramagneticendohedral metallofullerene and 1,2,3,4,5-pentamethylcyclopentadiene has been studied. three-step large-scale preparation of 1,2,3,4,5-pentamethylcyclopentadiene has been reported. reaction of 1,2,3,4,5-pentamethylcyclopentadiene (hcp*) with actinide ions in gas phase has been investigated by laser ablation mass spectrometry. it undergoes radical cation catalyzed cycloaddition with electron rich allenes to form diels-alder product.1,2,3,4,5-pentamethylcyclopentadiene was used as:
  • Growth modifier chemical, during metal organic chemical vapour deposition of iron from iron pentacarbonyl
  • Ligand in one-pot iridium-catalyzed transformation of alcohols to amides via the intermediacy of oximes
  • Raw material for the synthesis of [cp*rh(bpy)h2o]2+ (cp* = pentamethylcyclopentadienyl, bpy = 2,2α-bipyridyl), an electron mediator in the regeneration process of nadh

Specifications

CAS 4045-44-7
Density 0.87 g/mL (at 25°C (literature))
Boiling Point 58°C (13 mmHg)
Molecular Formula C10H16
Refractive Index n20/D 1.474 (literature)
Linear Formula C10H16
MDL Number MFCD00001354
Quantity 5 g
Molecular Weight (g/mol) 136.23
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.