Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4,5-Pentamethylcyclopentadiene, 95%

Catalog No. AC320830010
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC320830010 1 mL Glass bottle
AC320830050 5 mL Glass bottle
AC320830250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC320830010 Supplier Thermo Scientific Chemicals Supplier No. 320830010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 4045-44-7 | C10H16 | 136.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4045-44-7
Molecular Formula C10H16
Molecular Weight (g/mol) 136.11
MDL Number MFCD00001354
InChI Key WQIQNKQYEUMPBM-UHFFFAOYSA-N
Synonym 1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien
PubChem CID 77667
IUPAC Name 1,2,3,4,5-pentamethylcyclopenta-1,3-diene
SMILES CC1C(=C(C(=C1C)C)C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Orange-Yellow
Density 0.8700g/mL
Boiling Point 58°C (13.0 mmHg)
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4725 to 1.4755
Quantity 1 mL
Specific Gravity 0.87
Formula Weight 136.11
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1,2,3,4,5-Pentamethylcyclopentadiene, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 223-743-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.