Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,3,4,6-penta-O-acetyl-D-mannopyranose | 25941-03-1 | MFCD05864874 | 98.0% | 390.34 g/mol | C16H22O11 | 10 G
SDP

Catalog No. 5000301962
Encompass_Preferred

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is an acetyl-protected D-mannopyranose derivative used as a synthetic intermediate in carbohydrate chemistry and glycoside synthesis. Provided for research use, it is used to prepare acetylated mannopyranoside derivatives and other glycosylation intermediates.

  • Acetyl-protected sugar intermediate.
  • Purity 98.0%.
  • Molecular weight 390.34 g/mol.
  • Chemical formula C16H22O11.
  • White to off-white solid.
  • Soluble in DMSO (≈200 mg/mL; may require sonication).
  • Storage: powder -20°C (up to 3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Supplied in laboratory-scale quantities suitable for synthetic use.

Catalog No. 50-003-01962 Supplier Medchemexpress LLC Supplier No. HYW03989910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is an acetyl-protected D-mannopyranose derivative used as a synthetic intermediate in carbohydrate chemistry and glycoside synthesis. Provided for research use, it is used to prepare acetylated mannopyranoside derivatives and other glycosylation intermediates.

  • Acetyl-protected sugar intermediate.
  • Purity 98.0%.
  • Molecular weight 390.34 g/mol.
  • Chemical formula C16H22O11.
  • White to off-white solid.
  • Soluble in DMSO (≈200 mg/mL; may require sonication).
  • Storage: powder -20°C (up to 3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Supplied in laboratory-scale quantities suitable for synthetic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.