Learn More
Cayman Chemical 1 2 3 4 6-Penta-O-galoylD 10mg
SDP
Catalog No. 502968538
Encompass_Preferred
A major component of the root of P. suffruiticosa that at 30 µM inhibits the growth of human hepatocellular carcinoma SK-HEP-1 cells by causing cell cycle arrest and suppressing the activation of NF-κB
Catalog No. 50-296-8538 Supplier Cayman Chemical Supplier No. 1600710 MG
A major component of the root of P. suffruiticosa that at 30 µM inhibits the growth of human hepatocellular carcinoma SK-HEP-1 cells by causing cell cycle arrest and suppressing the activation of NF-κB