Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4,6-Penta-O-pivaloyl-D-mannopyranose 98.0+%, TCI America™
SDP

Catalog No. P18031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18031G Supplier TCI America Supplier No. P18031G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 220017-47-0
Molecular Formula C31H52O11
Molecular Weight (g/mol) 600.746
InChI Key PLXCBOJERHYNCS-AUGMSIGLSA-N
PubChem CID 11039291
IUPAC Name [(2R,3R,4S,5S)-3,4,5,6-tetrakis(2,2-dimethylpropanoyloxy)oxan-2-yl]methyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OCC1C(C(C(C(O1)OC(=O)C(C)(C)C)OC(=O)C(C)(C)C)OC(=O)C(C)(C)C)OC(=O)C(C)(C)C

Specifications

Melting Point 169°C
Color White
Quantity 1 g
Formula Weight 600.75
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4,6-Penta-O-pivaloyl-D-mannopyranose
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.