Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,3,4-Butanetetrol, (2R,3S)-rel- | 149-32-6 | 99.7% | 122.12 | C4H10O4 | 10 MG
SDP

Catalog No. 5000212350
Encompass_Preferred

meso-Erythritol is an analytical reference standard of meso-erythritol supplied for research and analytical applications. This sugar alcohol occurs naturally in fruits and is used as a low-calorie sweetener; as a certified standard it supports method development, calibration, and quality control in analytical laboratories.

  • High purity suitable for analytical use (99.73%).
  • Known chemical identity: 1,2,3,4-butanetetrol, (2R,3S)-rel-; CAS 149-32-6.
  • Molecular weight 122.12 g/mol and formula C4H10O4.
  • Supplied in small, accurately weighed quantities for traceable calibration.
  • Suitable for HPLC, GC, and other analytical method development and validation.

Catalog No. 50-002-12350 Supplier Medchemexpress LLC Supplier No. HY100551R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

meso-Erythritol is an analytical reference standard of meso-erythritol supplied for research and analytical applications. This sugar alcohol occurs naturally in fruits and is used as a low-calorie sweetener; as a certified standard it supports method development, calibration, and quality control in analytical laboratories.

  • High purity suitable for analytical use (99.73%).
  • Known chemical identity: 1,2,3,4-butanetetrol, (2R,3S)-rel-; CAS 149-32-6.
  • Molecular weight 122.12 g/mol and formula C4H10O4.
  • Supplied in small, accurately weighed quantities for traceable calibration.
  • Suitable for HPLC, GC, and other analytical method development and validation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.