Learn More
Medchemexpress LLC 1,2,3,4-butanetetrol, (2R,3S)-rel- | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g·mol^-1 | C4H10O4 | 500 MG
SDP
Catalog No. 5000212670
Encompass_Preferred
meso-Erythritol is an analytical-grade sugar alcohol used as a reference standard and research reagent. It occurs naturally in fruits such as pear and watermelon, has CAS number 149-32-6, molecular formula C4H10O4, and a molecular weight of 122.12 g·mol^-1. It is approximately 60-80% as sweet as sucrose and is used in studies of sweeteners, formulation research, and analytical method development.
  • Analytical standard suitable for research and quantitative assays.
  • High purity (99.7%) for reliable analytical results.
  • Well-characterized physicochemical properties (C4H10O4; 122.12 g·mol^-1).
  • Useful for sweetener and formulation studies due to its low-calorie sweetening profile.
  • Appropriate for use in analytical method development and validation.
Catalog No. 50-002-12670 Supplier Medchemexpress LLC Supplier No. HY100551500MG
meso-Erythritol is an analytical-grade sugar alcohol used as a reference standard and research reagent. It occurs naturally in fruits such as pear and watermelon, has CAS number 149-32-6, molecular formula C4H10O4, and a molecular weight of 122.12 g·mol^-1. It is approximately 60-80% as sweet as sucrose and is used in studies of sweeteners, formulation research, and analytical method development.
  • Analytical standard suitable for research and quantitative assays.
  • High purity (99.7%) for reliable analytical results.
  • Well-characterized physicochemical properties (C4H10O4; 122.12 g·mol^-1).
  • Useful for sweetener and formulation studies due to its low-calorie sweetening profile.
  • Appropriate for use in analytical method development and validation.